Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCATATGATCTGGATGTCAGATGC[C/T]GGAGTGTGGAAGCCATAGTGCACTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Hepacam2 PubMed Links |
Hepacam2 - HEPACAM family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178899.5 | 203 | Silent Mutation | NP_849230.1 |