Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCTCACAGTCCTTGAAGTAGGCG[A/G]TATTATTTCTGATTCCTACTATATA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Gab3 PubMed Links |
Gab3 - growth factor receptor bound protein 2-associated protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181584.4 | Intron | NP_853615.1 |