Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGCTACTCCTTAACAATCTTGTG[C/T]TCAGAATCATTCAAGCCTCTTGCCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Kcnk10 PubMed Links |
Kcnk10 - potassium channel, subfamily K, member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_029911.4 | Intron | NP_084187.2 |