Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACTATTCGCCGTGCTTCTGACAG[C/T]CCCTCTAATCCACTTCCCTGTAAGT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Slc38a6 PubMed Links |
Slc38a6 - solute carrier family 38, member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037717.3 | 1111 | Missense Mutation | NP_001032806.2 |