Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGACCTCATCGCTGAGGGAGACCG[A/C]GATGAGGGTGAGCTTCAGACTCGAC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Btbd7 PubMed Links |
Btbd7 - BTB (POZ) domain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172806.2 | Intron | NP_766394.2 |