Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGAGGCTAAAACCAAGCAGCTTCC[C/T]AAGTTCTTCCTCATAGAAGAAAAAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Trrap PubMed Links |
Trrap - transformation/transcription domain-associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081362.1 | 10449 | Silent Mutation | NP_001074831.1 |