Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Impa1 PubMed Links |
Impa1 - inositol (myo)-1(or 4)-monophosphatase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018864.5 | Intron | NP_061352.2 |
Slc10a5 - solute carrier family 10 (sodium/bile acid cotransporter family), member 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |