Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGGCGTGGGAGACTGAGGAGGA[A/G]ATGGGGTTATGGCCTGGGTGGATTC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Agps PubMed Links |
Agps - alkylglycerone phosphate synthase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172666.3 | Intron | NP_766254.2 |