Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGAAAATAATCTAACCCATACTA[G/T]CAAATTAGCTACATAATTAAAATTC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Parp8 PubMed Links |
Parp8 - poly (ADP-ribose) polymerase family, member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081009.1 | Intron | NP_001074478.1 |