Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTTGGTTGTTACTGCCTCAGAGA[C/T]GATTACCCACTGGGACAGAGATGTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Wipi2 PubMed Links |
Wipi2 - WD repeat domain, phosphoinositide interacting 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178398.4 | Intron | NP_848485.1 |