Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATCTCACTGTGAAGTCTGCCTA[A/G]CAAAGCAATCTCTTGCTTTTTGAAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Pou6f2 PubMed Links |
Pou6f2 - POU domain, class 6, transcription factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175006.2 | Intron | NP_778171.2 |