Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATTTTCTCAGTAAACAGGTTGTA[A/T]AGAGGATTTATGAAGGGTGCTATTT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Fbxo36 PubMed Links |
Fbxo36 - F-box protein 36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025386.3 | Intron | NP_079662.1 |