Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGTTCCTGTGTGGCCAGTGAGA[C/T]TTCCACAGGGACCTCAGTGGCCGCT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Kif26b PubMed Links |
Kif26b - kinesin family member 26B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161665.1 | 1565 | Missense Mutation | NP_001155137.1 |