Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCACCTCTAGAACAGGAGACATT[A/G]CCAACAACCATAGTTACCTCTGCAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Col26a1 PubMed Links |
Col26a1 - collagen, type XXVI, alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024474.2 | Intron | NP_077794.2 |