Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGCTGATGTCTAATATTTCATAA[G/T]CCTTTGAGATGTGTAACTTAATACT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Foxn3 PubMed Links |
Foxn3 - forkhead box N3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_183186.2 | Intron | NP_899009.2 |