Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTATGATTCCAGCCCTGACCTGG[A/G]ACATTCTTTTTTCTCTGAACCATCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Srgap1 PubMed Links |
Srgap1 - SLIT-ROBO Rho GTPase activating protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081037.2 | Intron | NP_001074506.1 | ||||
NM_001242411.1 | Intron | NP_001229340.1 |