Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAATTAAATTCTCCCCAGGCAAAC[A/C]GGCTTCCCTACCAGGCAGGTATTCC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Kcnh7 PubMed Links |
Kcnh7 - potassium voltage-gated channel, subfamily H (eag-related), member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133207.2 | Intron | NP_573470.2 |