Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGGTATAGGCCATGGGGTTCAG[G/T]CCTGTAGCAAAGGTGGGGGAGCTTA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Prex1 PubMed Links |
Prex1 - phosphatidylinositol-3,4,5-trisphosphate-dependent Rac exchange factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177782.3 | 3114 | Silent Mutation | NP_808450.2 |