Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATGCAGTAGGTTCTGGAACATTC[C/T]ACTGGTTTCCCTCAGCTGATACTCC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Amer3 PubMed Links |
Amer3 - APC membrane recruitment 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_213727.2 | 4148 | UTR 3 | NP_998892.1 |