Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCTGATTGTTAGTAATGACAGCA[A/G]TAACAGTTATGTTAATGATAAACAC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Lppr1 PubMed Links |
Lppr1 - lipid phosphate phosphatase-related protein type 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178756.4 | Intron | NP_848871.1 |