Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGATTCAGGACAGCGGCTCTATCA[C/T]TAGAAATGATTCACAGGGAGGCGTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Lef1 PubMed Links |
Lef1 - lymphoid enhancer binding factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276402.1 | Intron | NP_001263331.1 | ||||
NM_001276403.1 | Intron | NP_001263332.1 | ||||
NM_010703.4 | Intron | NP_034833.2 |