Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAATACAAATTAGATCTAAAGGTG[A/G]GAGAATAGGGGGAAAGGAGCTGAAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Pde1a PubMed Links |
Pde1a - phosphodiesterase 1A, calmodulin-dependent | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009978.1 | Intron | NP_001009978.1 | ||||
NM_001009979.1 | Intron | NP_001009979.1 | ||||
NM_001159582.1 | Intron | NP_001153054.1 | ||||
NM_016744.4 | Intron | NP_058024.2 |