Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAGCCTGTTATTCTATACATAGT[C/T]ATTTTAGAATCAAAGGCTGTTTAAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Fam198b PubMed Links |
Fam198b - family with sequence similarity 198, member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133187.3 | Intron | NP_573450.2 |