Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATAGCACCACCCAAAGAAGATAT[A/G]TTGACAATGCAAAAAAGACGCATAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Bmpr1b PubMed Links |
Bmpr1b - bone morphogenetic protein receptor, type 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277216.1 | Intron | NP_001264145.1 | ||||
NM_001277217.1 | Intron | NP_001264146.1 | ||||
NM_001277218.1 | Intron | NP_001264147.1 | ||||
NM_001277220.1 | Intron | NP_001264149.1 | ||||
NM_007560.4 | Intron | NP_031586.1 |