Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Thap2 PubMed Links |
Thap2 - THAP domain containing, apoptosis associated protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Zfc3h1 - zinc finger, C3H1-type containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033261.2 | Intron | NP_001028433.2 |