Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCGCCCTGGAACTGCTCCTCCAG[C/T]GCATCCACTCTTAGGTGGTCTGAGC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Tmprss3 PubMed Links |
Tmprss3 - transmembrane protease, serine 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163776.1 | 675 | Silent Mutation | NP_001157248.1 | |||
NM_080727.2 | 675 | Silent Mutation | NP_542765.2 |