Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGTTACCACTCTATGGCCTTGTG[A/G]GGAACATGGGTGATTTTATGCTAAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
4632428N05Rik PubMed Links |
4632428N05Rik - RIKEN cDNA 4632428N05 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Cdh23 - cadherin 23 (otocadherin) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252635.1 | Intron | NP_001239564.1 | ||||
NM_023370.3 | Intron | NP_075859.2 |