Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGGAACGAAATATCCAAGAGACA[C/T]ATCTTCTTTCCAGTGGTCAACATTA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Robo2 PubMed Links |
Robo2 - roundabout homolog 2 (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175549.4 | Intron | NP_780758.3 |