Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGCAAGCAAGGAGGTCTTCACTG[A/G]GACTGAGATGGAAAGAATTGGTGAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Mapre2 PubMed Links |
Mapre2 - microtubule-associated protein, RP/EB family, member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162941.1 | Intron | NP_001156413.1 | ||||
NM_001162942.1 | Intron | NP_001156414.1 | ||||
NM_153058.4 | Intron | NP_694698.3 |