Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Aspg PubMed Links |
Aspg - asparaginase homolog (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081169.1 | 410 | Silent Mutation | NP_001074638.1 |
Mir3073a - microRNA 3073a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Mir3073b - microRNA 3073b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |