Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Map6d1 PubMed Links |
Map6d1 - MAP6 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198599.2 | Intron | NP_941001.2 |
Yeats2 - YEATS domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |