Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGAGGTGCTGTGACTAAGCTTTT[A/C]TCAAAGCCTGTGAGCAGTCAGCATT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cngb3 PubMed Links |
Cngb3 - cyclic nucleotide gated channel beta 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013927.2 | Intron | NP_038955.1 |