Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGTGTTTACACTCTCTTTGTGGC[A/G]CCTACAAGAGGGATTTTCGACCCTC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Pde10a PubMed Links |
Pde10a - phosphodiesterase 10A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_011866.2 | Intron | NP_035996.2 |