Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTACAGCCTCTTAAGACATGCAG[C/T]CGTCCAATTAGGGTTGGCTTGTCAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Rad51ap2 PubMed Links |
Rad51ap2 - RAD51 associated protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001111118.1 | 2865 | Silent Mutation | NP_001104588.1 |