Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCACTGCAGCAGCAACTAGAGA[C/T]GATTCATCTTAGGAAAGACAAAGCG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Commd10 PubMed Links |
Commd10 - COMM domain containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178377.4 | 305 | Missense Mutation | NP_848464.1 |