Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGCTTCACCTGGAGGGTGCTGGG[C/T]GGTTCCCCGGCGTCGCTGCGCATCT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cbx4 PubMed Links |
Cbx4 - chromobox 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007625.2 | 1669 | Silent Mutation | NP_031651.2 |