Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTGGAAAAGAGGATGTGGGCCTC[A/G]GATGGATGCAGGATCTTCTTCAGAC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ngef PubMed Links |
Ngef - neuronal guanine nucleotide exchange factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001111314.1 | 1192 | Silent Mutation | NP_001104784.1 | |||
NM_019867.2 | 1192 | Silent Mutation | NP_063920.2 |