Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATATCCTGTTATTCAGGATATTAC[C/T]GTGATCTGAATAAATAATAATACTT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Col19a1 PubMed Links |
Col19a1 - collagen, type XIX, alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007733.2 | Intron | NP_031759.2 |