Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTCATCAGAAGATTAAAAATAAA[C/T]AAATAACAGAAAGTCAACTGTTTCC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cadps2 PubMed Links |
Cadps2 - Ca2+-dependent activator protein for secretion 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252105.1 | Intron | NP_001239034.1 | ||||
NM_001252106.1 | Intron | NP_001239035.1 | ||||
NM_001252107.1 | Intron | NP_001239036.1 | ||||
NM_001252108.1 | Intron | NP_001239037.1 | ||||
NM_001252109.1 | Intron | NP_001239038.1 | ||||
NM_001252110.1 | Intron | NP_001239039.1 | ||||
NM_153163.4 | Intron | NP_694803.3 |