Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATGGAAGTCTTCAGATAGTAAAC[C/T]CTAGACTTGTTCTACAATCTTTCCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Rab5a PubMed Links |
Rab5a - RAB5A, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025887.4 | Intron | NP_080163.1 |