Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATTCTTATTTCCTGGACCTTCTC[G/T]TCCCACTCTGAGCAGATTAGCTGAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Vav2 PubMed Links |
Vav2 - vav 2 oncogene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_009500.1 | Intron | NP_033526.1 |