Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACGACAAATAGAATTCTGGCCTC[C/G]TCTTGCTGCAGCAGTAACATACTGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Odf1 PubMed Links |
Odf1 - outer dense fiber of sperm tails 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008757.3 | 539 | Silent Mutation | NP_032783.2 |