Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Dlgap5 PubMed Links |
Dlgap5 - discs, large (Drosophila) homolog-associated protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144553.2 | 2253 | Silent Mutation | NP_653136.2 |
Lgals3 - lectin, galactose binding, soluble 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |