Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGTGCCAAGATCAAGAAAGCCAC[A/C]GGCATCAAGAACGGCCTGGCGCTGC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
2610027F03Rik PubMed Links |
2610027F03Rik - RIKEN cDNA 2610027F03 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
En1 - engrailed 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_010133.2 | 1659 | Silent Mutation | NP_034263.2 |