Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTCTTATGGGTTTTGATACATGA[A/G]TGGCAGCATTGACTACCTTTCTTTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Macrod2 PubMed Links |
Macrod2 - MACRO domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013802.3 | Intron | NP_001013824.2 | ||||
NM_028387.1 | Intron | NP_082663.1 |