Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTACTCATATGAGTCTTCCATGGT[A/G]CTGTGGTTGGAGGACCAGACATGGC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cd180 PubMed Links |
Cd180 - CD180 antigen | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008533.2 | 2295 | UTR 3 | NP_032559.2 |