Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTAGAATCAAACGGCCTCCTTGGT[C/T]GAAGACAAATCTGCCAGAGAGGACA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cald1 PubMed Links |
Cald1 - caldesmon 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145575.3 | Intron | NP_663550.1 |