Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAAAGCAGGTGACATGCTAAACA[A/G]CAGCCAAGTAACTGCCCTGGGTATG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Lhfpl3 PubMed Links |
Lhfpl3 - lipoma HMGIC fusion partner-like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081231.2 | Intron | NP_001074700.2 | ||||
NM_029990.1 | Intron | NP_084266.1 |