Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAGCAGCCAGGAAAAGTGACTGG[C/T]GAAGACCGACTGGGCTGGGGCAGTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Pop1 PubMed Links |
Pop1 - processing of precursor 1, ribonuclease P/MRP family, (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_026340.3 | 1931 | Silent Mutation | NP_080616.1 | |||
NM_152894.2 | 1931 | Silent Mutation | NP_690854.1 |