Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGGAACCACACAAAATGCTGCCC[C/T]ATTGGCTAGTTCAGAACAACTATTT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Galnt13 PubMed Links |
Galnt13 - UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173030.2 | Intron | NP_766618.2 |